Learn More
Bellco Glass, Inc. Oven Rocker Tray
SDP
Catalog No. 501946531
Encompass_Preferred
Stainless steel rocker platform fit Bellco micro and mini ovens. Platform has 3 settings at 2.5 degree to 6 degrees from horizontal and adapts to existing rotisserie hardware. MADE IN THE USA
Catalog No. 50-194-6531 Supplier Bellco Glass, Inc. Supplier No. 793050100
Stainless steel rocker platform fit Bellco micro and mini ovens. Platform has 3 settings at 2.5 degree to 6 degrees from horizontal and adapts to existing rotisserie hardware. MADE IN THE USA