Learn More
VWR INTERNATIONAL INC Oven Thermometer (with 30 mL bottle of Glass Beads)
Catalog No. NC1601678
Encompass_Preferred
Range: +20 to 130°C, Length: 145 mm, Accuracy: ±1.5°C; ±2°C above 100°C, Divisions: 1°C
Catalog No. NC1601678 Supplier VWR INTERNATIONAL INC Supplier No. 89095690
Range: +20 to 130°C, Length: 145 mm, Accuracy: ±1.5°C; ±2°C above 100°C, Divisions: 1°C