Learn More
Thermco Products Inc Oven Thermometer +35 to 180C, 145mm, Sand - 100 ml Glass Bottle
Catalog No. NC1030112
Encompass_Preferred
Non-Mercury thermometers sealed in a PTEF jacket that contains liquid & glass if breakage occurs and are housed in unbreakable plastic bottles filled with a non-toxic Bio-Safe mediun. Economical, Precise and Reliable means of measuring critical temperatures in Ovens. Each bottle thermometer have their own unique serial number and are supplied with a NIST Traceable Certificate.
Catalog No. NC1030112 Supplier Thermco Products Inc Supplier No. ACCOV080S
Non-Mercury thermometers sealed in a PTEF jacket that contains liquid & glass if breakage occurs and are housed in unbreakable plastic bottles filled with a non-toxic Bio-Safe mediun. Economical, Precise and Reliable means of measuring critical temperatures in Ovens. Each bottle thermometer have their own unique serial number and are supplied with a NIST Traceable Certificate.