Learn More
Thermco Products Inc OVEN THRMMTR +35/200 MERCURY
Catalog No. NC1246326
Encompass_Preferred
Mercury thermometers sealed in a PTEF jacket that contains liquid & glass if breakage occurs and are housed in unbreakable plastic bottles filled with a non-toxic Bio-Safe mediun. Economical, Precise and Reliable means of measuring critical temperatures in Ovens. Each bottle thermometer have their own unique serial number and are supplied with a NIST Traceable Certificate.
Catalog No. NC1246326 Supplier Thermco Products Inc Supplier No. ACCOV080
Mercury thermometers sealed in a PTEF jacket that contains liquid & glass if breakage occurs and are housed in unbreakable plastic bottles filled with a non-toxic Bio-Safe mediun. Economical, Precise and Reliable means of measuring critical temperatures in Ovens. Each bottle thermometer have their own unique serial number and are supplied with a NIST Traceable Certificate.