Learn More
Chemglass Life Sciences 0.6 Cu. Ft. Oven, Vacuum, 120V
SDP
Catalog No. 50143554
Encompass
The Vacuum ovens are durably constructed with seamless welds and strong bracing. All of the vacuum ovens are built with a stainless steel chamber for exceptional durability. Ovens incorporate solid brass vacuum valves with PTFE seats to prevent leaks. The double plenum design exceeds CE safety requirements resulting in a cool outer surface. The doors on these units have positive latch handles with spring-loaded glass to facilitate a good vacuum seal without hinge binds that shorten the gasket life. A selection of gaskets (for specific applications) and a small footprint increase the versatility of these ovens. The unique cross-flow ventilation forces inert gas to fill the entire chamber. Users can choose from 3/8 inch orifices or a KF25 fitting to withstand heavy use, minimize draw-down time and achieve low vacuum levels. We offer a full line of gaskets to suit applications involving high temperatures, acids, or solvents. Ovens have a 24 month limited warranty.
Catalog No. 50-143-554 Supplier Chemglass Life Sciences Supplier No. CLS3450006
Add to Cart
Add to Cart
Request a Quote
The Vacuum ovens are durably constructed with seamless welds and strong bracing. All of the vacuum ovens are built with a stainless steel chamber for exceptional durability. Ovens incorporate solid brass vacuum valves with PTFE seats to prevent leaks. The double plenum design exceeds CE safety requirements resulting in a cool outer surface. The doors on these units have positive latch handles with spring-loaded glass to facilitate a good vacuum seal without hinge binds that shorten the gasket life. A selection of gaskets (for specific applications) and a small footprint increase the versatility of these ovens. The unique cross-flow ventilation forces inert gas to fill the entire chamber. Users can choose from 3/8 inch orifices or a KF25 fitting to withstand heavy use, minimize draw-down time and achieve low vacuum levels. We offer a full line of gaskets to suit applications involving high temperatures, acids, or solvents. Ovens have a 24 month limited warranty.