Learn More
METTLER TOLEDO 3P INDUSTRIAL Water Standard Oven 1%, 5 g
Catalog No. NC1818749
Encompass
Water standard oven 1% is used as certified reference material to check the accuracy of coulometric and volumetric water determinations.
Catalog No. NC1818749 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 63053004
Water standard oven 1% is used as certified reference material to check the accuracy of coulometric and volumetric water determinations.