Learn More
MSC ZEP Oven and Grill Cleaner, Lemon Scent, Gel, 4 ea.
Catalog No. 502817622
Encompass
This oven and grill cleaner features a lemon scent and is provided in a gel formulation. It is supplied in 1 gal. bottles, with four bottles per case, for efficient cleaning of commercial kitchen equipment.
  • Lemon scent
  • Gel formulation for effective cleaning
  • Supplied in 1 gal. bottles
  • Packaged as a case of 4 bottles
  • High pH range of 13 to 13.9
  • Contains sodium hydroxide at 10-20% concentration
  • Causes severe skin burns and eye damage
Catalog No. 50-281-7622 Supplier MSC Supplier No. 17595158
This oven and grill cleaner features a lemon scent and is provided in a gel formulation. It is supplied in 1 gal. bottles, with four bottles per case, for efficient cleaning of commercial kitchen equipment.
  • Lemon scent
  • Gel formulation for effective cleaning
  • Supplied in 1 gal. bottles
  • Packaged as a case of 4 bottles
  • High pH range of 13 to 13.9
  • Contains sodium hydroxide at 10-20% concentration
  • Causes severe skin burns and eye damage