Learn More
Electron Microscopy Sciences Stainless Steel Over Chucks Freezing Blocks
SDP
Catalog No. 50364891
Encompass_Preferred
Stainless steel rectangular blocks serve as a heat extractor, designed to fit over the stem of the freezing chuck. The flat surfaces can also be used as an extra heat extractor when using conventional cryostat embedding technique. The freezing block also served as a dislodging tool. A light tap of the chuck stem cleaves the plane of adhesion holding of the formed block to the well. These blocks also serve as a convenient hand held flat freezing surface useful in gently cooling the tissue without the use of sprays and in plastering technique.
Catalog No. 50-364-891 Supplier Electron Microscopy Sciences Supplier No. 6172501
Stainless steel rectangular blocks serve as a heat extractor, designed to fit over the stem of the freezing chuck. The flat surfaces can also be used as an extra heat extractor when using conventional cryostat embedding technique. The freezing block also served as a dislodging tool. A light tap of the chuck stem cleaves the plane of adhesion holding of the formed block to the well. These blocks also serve as a convenient hand held flat freezing surface useful in gently cooling the tissue without the use of sprays and in plastering technique.