Learn More
PATHOLOGY INNOVATIONS OVER CHUCK FREEZING BLOCK
Catalog No. NC0754413
Encompass_Preferred
These steel rectangular blocks are a tool useful for a number of tasks. They serve as a heat extractor, designed to fit over the stem of the chuck .The flat surfaces can also be used as an extra heat extractor when using conventional cryostat embedding technique. The freezing block also served as a dislodging tool. A light tap of the chuck stem cleaves the plane of adhesion holding of the formed block to the well. These blocks also serve as a convenient hand held flat freezing surface useful in gently cooling the tissue without the use of sprays and in plastering technique discussed below.
Catalog No. NC0754413 Supplier PATHOLOGY INNOVATIONS Supplier No. 141
These steel rectangular blocks are a tool useful for a number of tasks. They serve as a heat extractor, designed to fit over the stem of the chuck .The flat surfaces can also be used as an extra heat extractor when using conventional cryostat embedding technique. The freezing block also served as a dislodging tool. A light tap of the chuck stem cleaves the plane of adhesion holding of the formed block to the well. These blocks also serve as a convenient hand held flat freezing surface useful in gently cooling the tissue without the use of sprays and in plastering technique discussed below.