Learn More
LGC GENOMICS LLC (Lucigen) OverExpress C41(DE3) Electrocompetent Cells, 12 Rxns (SOLOs)
Supplier:  LGC GENOMICS LLC 603411
Encompass
OverExpress C41(DE3) Electrocompetent Cells, 12 Rxns (SOLOs)Express difficult or toxic proteins using standard E. coli T7 expression vectors using these unique strains. Also available in chemically competent format.Express genes cloned into any T7 vector with these BL21(DE3) derivativesEffective in expressing toxic membrane proteinsCited in over 350 research articlesRecommended for membrane proteins with T7 vectorEach OverExpress kit contains Electrocompetent Cells in SOLO packaging (1 transformation per tube), Expression Recovery Medium (lactose minus), pUC19 Positive Control Plasmid, pAVD10 Verification Plasmid, and complete protocols.
Catalog No. NC2160375