Learn More
Chemglass Life Sciences Overhead Drive Adapter, Standard, 161mm OAH, 78mm from cap to top
SDP
Catalog No. 50143187
Encompass
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.
Catalog No. 50-143-187 Supplier Chemglass Life Sciences Supplier No. CLS144010
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.