Learn More
Research Products International Corp Overhead Stirrer for Tissue Homogenizing
SDP
Catalog No. 501976904
Encompass
This compact stirrer delivers the necessary speed and power for tissue homogenizing, tissue, stirrer, mixing, homogenizing, grinders. Wheaton Industries Inc. electronic control allows precise selection of speeds from 1,000-10,000 RPM. Designed for use with tissue grinders having a steel rod and PTFE pestle. Support stand not included.
Catalog No. 50-197-6904 Supplier Research Products International Corp Supplier No. 903475
This compact stirrer delivers the necessary speed and power for tissue homogenizing, tissue, stirrer, mixing, homogenizing, grinders. Wheaton Industries Inc. electronic control allows precise selection of speeds from 1,000-10,000 RPM. Designed for use with tissue grinders having a steel rod and PTFE pestle. Support stand not included.