Learn More
BINDER™ Overheating Alarm
SDP
Catalog No. 08111806 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-806 Plastic
1 options
Catalog No. 08-111-806 Supplier BINDER™ Supplier No. 80121636
Add to Cart
Add to Cart
For Use With: FD 56, FD 115, FD 260, FD 720
  • Audible
  • Can be switched off
  • Adjustable limit on the independent temperature safety device
  • Switchable acoustic alarm
Specifications
Description Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
For Use With (Equipment) BINDER Chambers FD 56, FD 115, FD 260, FD 720
Material Plastic
Type Alarm