Learn More
BINDER™ Overheating Alarm
SDP
Catalog No. 08111568 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BF 400
Catalog No. For Use With (Equipment)
08-111-568 BF 400
1 options
Catalog No. 08-111-568 Supplier BINDER™ Supplier No. 80121302
Add to Cart
Add to Cart
For Use With: BF 400
  • Audible
  • Can be switched off
  • Adjustable limit on the independent temperature safety device
  • Switchable acoustic alarm
Specifications
For Use With (Equipment) BF 400
Material Plastic
Type Overheating Alarm
Voltage 120 V