Learn More
BINDER™ Overheating Alarm
SDP
Catalog No. 08111569 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-569 Plastic
1 options
Catalog No. 08-111-569 Supplier BINDER™ Supplier No. 80121303
Add to Cart
Add to Cart
For Use With: ED 23, ED 400
  • Audible
  • Can be switched off
  • Adjustable limit on the independent temperature safety device
  • Switchable acoustic alarm
Specifications
For Use With (Equipment) ED 23, ED 400
Material Plastic
Type Overheating Alarm
Voltage 120/208 V