Learn More
Chemglass Life Sciences 5" Overlay Gas Assembly, L/S Platinum Cured Silicone Pump Tubing, 1/8" ID x 1/4" OD Size 16
SDP
Catalog No. 5000083232
Encompass
Features/Benefits: Single Use: Minimize Labor CostsEliminates Cleaning ValidationReduces Set up Time and Costs.Pre-Fabricated Assemblies: Minimize Injuries associated with cutting in-houseImproved consistency.Offers Closed System: Gives the ability to assemble in Open Lab type EnvironmentProvides aseptic solution to close off an unused line.How to Sterilize: Autoclave Max 15 psi @ 134 degreesC, 60 minGamma Radiation 25 kGy to 50 kGy.
Catalog No. 50-000-83232 Supplier Chemglass Life Sciences Supplier No. CLS139014
Features/Benefits: Single Use: Minimize Labor CostsEliminates Cleaning ValidationReduces Set up Time and Costs.Pre-Fabricated Assemblies: Minimize Injuries associated with cutting in-houseImproved consistency.Offers Closed System: Gives the ability to assemble in Open Lab type EnvironmentProvides aseptic solution to close off an unused line.How to Sterilize: Autoclave Max 15 psi @ 134 degreesC, 60 minGamma Radiation 25 kGy to 50 kGy.