Learn More
Grainger Schneider Electric Industrial Relay, 1.6 to 2.5 A, Class 20, 3-Pole, 1 ea
Catalog No. 502795408
Encompass_Preferred
The TeSys Deca thermal overload relay is a 3-pole, bimetallic device engineered for direct mounting onto TeSys D contactors, specifically designed for industrial motor protection. It features a thermal setting range of 1.6 A to 2.5 A and is rated for tripping class 20, providing reliable operation for motors up to 0.75 kW at 400 V. This compact, temperature-compensated relay includes integrated phase unbalance and phase failure detection to ensure motor longevity in demanding applications.
  • Tripping class 20 for advanced motor protection
  • Integrated phase unbalance and phase failure detection
  • Manual and automatic reset options via a selectable switch
  • Includes 1NO and 1NC auxiliary contacts for trip signaling
  • Features red test and blue reset/stop pushbuttons
  • Compatible with contactors from 9 A to 38 A
  • IP20 degree of protection against direct finger contact
Catalog No. 50-279-5408 Supplier Grainger Supplier No. 19YD03
The TeSys Deca thermal overload relay is a 3-pole, bimetallic device engineered for direct mounting onto TeSys D contactors, specifically designed for industrial motor protection. It features a thermal setting range of 1.6 A to 2.5 A and is rated for tripping class 20, providing reliable operation for motors up to 0.75 kW at 400 V. This compact, temperature-compensated relay includes integrated phase unbalance and phase failure detection to ensure motor longevity in demanding applications.
  • Tripping class 20 for advanced motor protection
  • Integrated phase unbalance and phase failure detection
  • Manual and automatic reset options via a selectable switch
  • Includes 1NO and 1NC auxiliary contacts for trip signaling
  • Features red test and blue reset/stop pushbuttons
  • Compatible with contactors from 9 A to 38 A
  • IP20 degree of protection against direct finger contact