Learn More
Grainger Schneider Electric TeSys Deca Thermal Overload Relay, 12-18 A, Class 20, Bimetallic, 1 ea
Catalog No. 502796085
Encompass_Preferred
This bimetallic thermal overload relay is engineered for reliable motor protection in industrial applications. Designed for direct mounting on specified contactors to form a compact starter, it provides precise monitoring for motors up to 7.5 kW at 400 V. This device features a thermal protection adjustment range of 12 A to 18 A and a class 20 tripping characteristic.
  • Features a thermal adjustment range of 12 A to 18 A and temperature compensation for high accuracy.
  • Includes manual test and reset buttons, an automatic reset selector, and a trip flag indicator.
  • Equipped with 1NO and 1NC auxiliary contacts for trip signaling.
  • Supports direct mounting on compatible contactors, with options for separate mounting or remote control accessories.
  • Certified to international standards including IEC, UL, CSA, CCC, EAC, and Marine ratings.
Catalog No. 50-279-6085 Supplier Grainger Supplier No. 19YC96
This bimetallic thermal overload relay is engineered for reliable motor protection in industrial applications. Designed for direct mounting on specified contactors to form a compact starter, it provides precise monitoring for motors up to 7.5 kW at 400 V. This device features a thermal protection adjustment range of 12 A to 18 A and a class 20 tripping characteristic.
  • Features a thermal adjustment range of 12 A to 18 A and temperature compensation for high accuracy.
  • Includes manual test and reset buttons, an automatic reset selector, and a trip flag indicator.
  • Equipped with 1NO and 1NC auxiliary contacts for trip signaling.
  • Supports direct mounting on compatible contactors, with options for separate mounting or remote control accessories.
  • Certified to international standards including IEC, UL, CSA, CCC, EAC, and Marine ratings.