Learn More
Grainger Schneider Electric TeSys Deca Thermal Overload Relay, 2.5-4 A, Class 20, 1 EA
Catalog No. 502795406
Encompass_Preferred
The TeSys Deca is a non-differential thermal overload relay designed for reliable motor protection. This bimetallic relay operates within a 2.5 A to 4 A range and is specifically engineered for use with unbalanced loads. Featuring a class 20 trip rating, it provides precise protection for motors and integrates seamlessly into motor control systems.
  • Thermal protection adjustment range of 2.5 A to 4 A
  • Class 20 tripping class for motor protection
  • Equipped with 1 NO + 1 NC auxiliary contacts for trip signaling
  • Manual and automatic reset selector with trip flag indicator
  • Temperature compensated for consistent performance from -20°C to 60°C
  • Designed for direct mounting on the bottom terminals of LC1D09-D38 contactors
  • Rated operational voltage of 690 V AC (0 to 400 Hz)
  • IP20 degree of protection
Catalog No. 50-279-5406 Supplier Grainger Supplier No. 19YC91
The TeSys Deca is a non-differential thermal overload relay designed for reliable motor protection. This bimetallic relay operates within a 2.5 A to 4 A range and is specifically engineered for use with unbalanced loads. Featuring a class 20 trip rating, it provides precise protection for motors and integrates seamlessly into motor control systems.
  • Thermal protection adjustment range of 2.5 A to 4 A
  • Class 20 tripping class for motor protection
  • Equipped with 1 NO + 1 NC auxiliary contacts for trip signaling
  • Manual and automatic reset selector with trip flag indicator
  • Temperature compensated for consistent performance from -20°C to 60°C
  • Designed for direct mounting on the bottom terminals of LC1D09-D38 contactors
  • Rated operational voltage of 690 V AC (0 to 400 Hz)
  • IP20 degree of protection