Learn More
Grainger TeSys Deca Thermal Overload Relay, 23 to 32 A, Class 20, 3-Pole, 1 ea
Catalog No. 502795684
Encompass_Preferred
The TeSys Deca is a 3-pole, non-differential thermal overload relay designed for reliable motor protection. Engineered for direct mounting to compatible contactors, this bimetallic relay features an adjustable thermal protection range of 23 A to 32 A and operates across a network frequency range of 0 to 400 Hz. It offers robust performance with an IP20 degree of protection.
  • Trip class 20 for effective motor overload protection.
  • Features phase failure sensitivity where tripping occurs at 130% of Ir on two phases.
  • Integrated 1 NO and 1 NC auxiliary contacts with a trip flag indicator.
  • Dual control options include a red push-button for stop and a blue push-button for manual or automatic reset.
  • Temperature compensated for operation between -20°C and 60°C (-4°F to 140°F).
  • Screw clamp terminals for secure power and control circuit installation.
Catalog No. 50-279-5684 Supplier Grainger Supplier No. 19YC98
The TeSys Deca is a 3-pole, non-differential thermal overload relay designed for reliable motor protection. Engineered for direct mounting to compatible contactors, this bimetallic relay features an adjustable thermal protection range of 23 A to 32 A and operates across a network frequency range of 0 to 400 Hz. It offers robust performance with an IP20 degree of protection.
  • Trip class 20 for effective motor overload protection.
  • Features phase failure sensitivity where tripping occurs at 130% of Ir on two phases.
  • Integrated 1 NO and 1 NC auxiliary contacts with a trip flag indicator.
  • Dual control options include a red push-button for stop and a blue push-button for manual or automatic reset.
  • Temperature compensated for operation between -20°C and 60°C (-4°F to 140°F).
  • Screw clamp terminals for secure power and control circuit installation.