Learn More
Grainger Schneider Electric TeSys Deca Thermal Overload Relay, 5.5 to 8 A, Class 20, 1 ea
Catalog No. 502795180
Encompass_Preferred
This bimetallic thermal overload relay provides reliable motor protection with a thermal setting range of 5.5 to 8 A and class 20 tripping. Designed for integration with contactors, this unit includes phase unbalance detection and temperature compensation to ensure accurate motor operation. It features a compact design for space-saving installations and supports versatile manual or automatic reset configurations.
  • 5.5 to 8 A thermal adjustment range
  • Class 20 tripping
  • Phase unbalance detection
  • Temperature compensated for protection accuracy
  • Manual test and reset buttons with manual/automatic reset selector
  • 1NO+1NC auxiliary contacts and trip flag indicator
  • Compatible with direct mounting on contactors for compact starter assembly
  • Multi-standards certified
Catalog No. 50-279-5180 Supplier Grainger Supplier No. 19YD06
This bimetallic thermal overload relay provides reliable motor protection with a thermal setting range of 5.5 to 8 A and class 20 tripping. Designed for integration with contactors, this unit includes phase unbalance detection and temperature compensation to ensure accurate motor operation. It features a compact design for space-saving installations and supports versatile manual or automatic reset configurations.
  • 5.5 to 8 A thermal adjustment range
  • Class 20 tripping
  • Phase unbalance detection
  • Temperature compensated for protection accuracy
  • Manual test and reset buttons with manual/automatic reset selector
  • 1NO+1NC auxiliary contacts and trip flag indicator
  • Compatible with direct mounting on contactors for compact starter assembly
  • Multi-standards certified