Learn More
Grainger Overnight Hygiene Admission Kit PK100
Catalog No. 502753597
Encompass_Preferred
The Grainger Approved PK-100 is a 100-foot extension cord that provides temporary power where needed. It features a 16 gauge, 3-wire SJT cord with a grounded plug on one end and a grounded receptacle on the other. The flexible vinyl jacket resists oil and water. This general purpose extension cord is UL listed and CSA certified. It can handle up to 15 amps and 1875 watts. The PK-100 is ideal for both indoor and outdoor applications.
Catalog No. 50-275-3597 Supplier Grainger Supplier No. 13Z980
The Grainger Approved PK-100 is a 100-foot extension cord that provides temporary power where needed. It features a 16 gauge, 3-wire SJT cord with a grounded plug on one end and a grounded receptacle on the other. The flexible vinyl jacket resists oil and water. This general purpose extension cord is UL listed and CSA certified. It can handle up to 15 amps and 1875 watts. The PK-100 is ideal for both indoor and outdoor applications.