Learn More
Labelmaster Inc 'OVERPACK' DOT LABEL RED ON WH
SDP
Catalog No. NC0114695
Encompass_Preferred
Overpacks containing dangerous goods are required to be marked with the word, Overpack, in association with the proper shipping name. This rule is effective for international air and sea shipments; is required for shipment in the U.S.
Catalog No. NC0114695 Supplier Labelmaster Inc Supplier No. L370
Overpacks containing dangerous goods are required to be marked with the word, Overpack, in association with the proper shipping name. This rule is effective for international air and sea shipments; is required for shipment in the U.S.