Learn More
Saf T Pak Inc STP-510-OVPK Category B and Exempt Patient Specimen, Ambient Insulated Shipping System, (UN 3373 and Exempt) Medium, 10/Case
SDP
Catalog No. 501904611 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
Overpack means an enclosure that is used by a single consignor to provide refrigeration, protection or convenience in the handling of a single fully compliant package or to consolidate two or more fully compliant packages.FEATURES:Use with two STP-309SYS for " Dual Shipper" - two temperature requirements in one shipmentSystem Components:Outer Box OD 19.75 x 12.5 x 9.5", pre-printed with Overpack MarkingSTP-803 Class 9 Hazard Label (1 ea.)STP-804 UN 1845 " Dry ice Quantity" Marking Label (1 ea.)STP-818 UN 3373 Marking Label (1 ea.)STP-818 UN 3373 Marking Label (1 ea.)STP-819 Exempt Human/Animal Specimen Label (1 ea.)STP-820 Biological Substance, Category B Marking (1 ea.)
Catalog No. 50-190-4611 Supplier Saf T Pak Inc Supplier No. 35183
Overpack means an enclosure that is used by a single consignor to provide refrigeration, protection or convenience in the handling of a single fully compliant package or to consolidate two or more fully compliant packages.FEATURES:Use with two STP-309SYS for " Dual Shipper" - two temperature requirements in one shipmentSystem Components:Outer Box OD 19.75 x 12.5 x 9.5", pre-printed with Overpack MarkingSTP-803 Class 9 Hazard Label (1 ea.)STP-804 UN 1845 " Dry ice Quantity" Marking Label (1 ea.)STP-818 UN 3373 Marking Label (1 ea.)STP-818 UN 3373 Marking Label (1 ea.)STP-819 Exempt Human/Animal Specimen Label (1 ea.)STP-820 Biological Substance, Category B Marking (1 ea.)