Learn More
Cole-Parmer Cole-Parmer OVF-800-147-120 Mechanical Convection Oven, 147 L; 120 VAC
Catalog No. NC2675859 Shop All Cole-Parmer Products
Encompass_Preferred
Optimized airflow provides even heat transfer for high temperature uniformityThree-point temperature calibration ensures excellent temperature accuracy over a wide temperature rangeIntuitive 3.5” touch screen controllerStainless steel interior resists corrosionWide-latch handle makes it easy to open and close door even when hands are fullStay-cool exterior for user safety
Catalog No. NC2675859 Supplier Cole-Parmer Supplier No. 5241222
Optimized airflow provides even heat transfer for high temperature uniformityThree-point temperature calibration ensures excellent temperature accuracy over a wide temperature rangeIntuitive 3.5” touch screen controllerStainless steel interior resists corrosionWide-latch handle makes it easy to open and close door even when hands are fullStay-cool exterior for user safety