Learn More
Raybiotech Inc G-Series Ovine Cytokine Array 2 Kit.
Catalog No. 501950480
Encompass_Preferred
Detects 10 Ovine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: AIF, Cadherin-6, IFN alpha, IL-1 alpha, IL-1 beta, IL-4, IL-13, LEKTI, LIF, RANTES (CCL5)
Catalog No. 50-195-0480 Supplier Raybiotech Inc Supplier No. GSOCYT21
Detects 10 Ovine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: AIF, Cadherin-6, IFN alpha, IL-1 alpha, IL-1 beta, IL-4, IL-13, LEKTI, LIF, RANTES (CCL5)