Learn More
Raybiotech Inc G-Series Ovine Cytokine Array 20 Kit.
Catalog No. 501950477
Encompass_Preferred
Semi-Quantitative measurement of 20 Ovine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 6Ckine, AMSH, Angiopoietin-1, ANGPTL3, BLC, BLMH, Complement MASP3, Contactin-1, FLRG, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-13, IL-17A, IL-17F, IL-21, IL-8, Leptin, Matrilin-3, MIP-1 beta, MMP-9, NCAM-1, Notch-3, RANTES, SCF, SREC-II, Thrombospondin-2, TNF alpha, TWEAK R, VEGF-A
Catalog No. 50-195-0477 Supplier Raybiotech Inc Supplier No. GSOCAA202
Semi-Quantitative measurement of 20 Ovine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 6Ckine, AMSH, Angiopoietin-1, ANGPTL3, BLC, BLMH, Complement MASP3, Contactin-1, FLRG, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-13, IL-17A, IL-17F, IL-21, IL-8, Leptin, Matrilin-3, MIP-1 beta, MMP-9, NCAM-1, Notch-3, RANTES, SCF, SREC-II, Thrombospondin-2, TNF alpha, TWEAK R, VEGF-A