Learn More
Kingfisher Biotech Inc Ovine IFN gamma Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0491V005
Encompass
The Ovine IFN gamma yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Ovine IFN gamma applications are for cell culture, ELISA standard, and Western Blot Control. The Ovine IFN gamma yeast-derived recombinant protein can be purchased in multiple sizes. Ovine IFN gamma Specifications: (Molecular Weight: 16.9 kDa) (Amino Acid Sequence: QGPFFKEIEN LKEYFNASNP DVAKGGPLFS EILKNWKEES DKKIIQSQIV SFYFKLFENL KDNQVIQRSM DIIKQDMFQK FLNGSSEKLE DFKRLIQIPV DDLQIQRKAI NELIKVMNDL SPKSNLRKRK RSQNLFRGRR ASM (143)) (Gene ID: 443396). For research use only. Made in the USA
Catalog No. 50-241-3927