Learn More
Raybiotech Inc Ovine IL-1 beta/IL-1 F2 ELISA Kit
Supplier:  Raybiotech Inc ELOIL1B1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 13 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Ovine IL-1 beta.
Catalog No. 50-195-0170