Learn More
Raybiotech Inc Ovine IP-10 (CXCL10) ELISA Kit
Supplier:  Raybiotech Inc ELOIP101
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 28 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Ovine MIG, Decorin, IFN-gamma, IL-8, IL-17A, IL-21, sFRP-3, TNF-alpha, VEGF-A
Catalog No. 50-195-0173