Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ovine Testes

Catalog No. mp215127425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215127425 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP215127425 Supplier MP Biomedicals™ Supplier No. 0215127425
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in conjunction with collagenase to dissociate the extracellular matrix between cells of animal tissue, in order to release viable cells for use in tissue culture.

Applications: Testicular hyaluronidase hydrolyzes the endo-N-acetylhexosaminic bonds of hyaluronic acid and chondroitin sulfuric acids A and C (but not B), primarily to tetrasaccharide residues. Monosaccharides are not liberated. Hyaluronidase is often used in conjunction with collagenase to dissociate the extracellular matrix between cells of animal tissue, in order to release viable cells for use in tissue culture. It may also be used to clarify synovial fluids in order to make cell counts possible.
Product Description: Hyaluronidase is a glycoprotein containing 5% mannose and 2.17% glucosamine, it catalyzes the random hydrolysis of 1,4-linkages between 2-acetamido- 2-deoxy- β-D-glucose and D-glucose residues in hyaluronate. Hyaluronidase from bovine testes is a tetramer consisting of 4 equal subunits with a molecular mass of 14 kDa each.
Biochem/physiol Actions: Hyaluronidase degrades hyaluronan and has been found to be inappropriately regulated during cancer progression. These enzymes randomly cleave β-N-acetylhexosamine-[1→4] glycosidic bonds in hyaluronic acid, chondroitin, and chondroitin sulfates.
Unit Definition: Amount of enzyme which liberates one micromole of N-acetylglucosamine per minute at 37°C and pH 4.0
Specificity: Testicular hyaluronidase hydrolyzes the endo-N-acetylhexosaminic bonds of hyaluronic acid and chondroitin sulfuric acids A and C (but not B), primarily to tetrasaccharide residues. Monosaccharides are not liberated.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Cell Dissociation
Formula Weight ∽55 kDa (four subunits of 14 kDa each)
Synonym Hyaluronate 4-glycanohydrolase, hyaluronate lyase, hyaluronoglucosaminidase, 3.2.1.35
Solubility Dissolve 10 mg hyaluronic acid in 25 mL 0.1 M sodium phosphate buffer: pH 5.3 with 0.15 M sodium chloride.
Content And Storage Store at -20°C and dessicate
Form Lyophilized
For Use With (Application) Drug dispersion & delivery
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.