Learn More
Aviva Systems Biology Corp OVOL1 Antibody - C-terminal region (ARP38500_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38500_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against OVOL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human OVOL1. Alternate names: HOVO1ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56677