Learn More
Aviva Systems Biology Corp OVOL2 Antibody - middle region (ARP39500_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39500_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OVOL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OVOL2. Alternate names: EUROIMAGE566589, ZNF339ApplicationWB, IF. Species reactivity: Human
Catalog No. 50-014-57059