Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Owen Mumford™ Lancet Kit
SDP

Catalog No. 22024003 Shop All Owen Mumford, Inc. Products
Change view
Click to view available options
Quantity:
100 Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-024-003 100 Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-024-003 Supplier Owen Mumford™ Supplier No. NT 0100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

One 10g monofilament and one Neurotip

Dual-purpose, pocket-size neuropathy screening device provides a safe and reliable force

  • Neuropen assesses if patient can distinguish touch stimulus (monofilament) and sharp stimulus (Neurotip)
  • Neurotip test uses calibrated spring mechanism to force of 40g to assess reduced sensation to sharpness/pain in small nerve fibers
  • Monofilament test exerts force of 10g to assess protective touch/pressure sensation in large nerve fibers
  • Monofilaments are replaceable

Peripheral neuropathy cannot be excluded without regular foot screening in conjunction with symptomatic and clinical assessments

Specifications

For Use With (Application) For foot screening
Product Type Lancet Kit
Quantity 100 Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.