Learn More
Owen Mumford™ Unistik™ 3 Gentle Single use Safety Lancets
SDP
Catalog No. 22023390 Shop All Owen Mumford, Inc. Products
Change view
Click to view available options
Packaging Quantity:
100/Pk.
200/Pk.
25/Pk.
50/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
22-023-390 25/Pk.
22-023-388 100/Pk.
22-023-387 200/Pk.
22-023-389 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-023-390 Supplier Owen Mumford™ Supplier No. AT 1028
Add to Cart
Add to Cart
This item is not returnable. View return policy
For pediatric and frequent testing
  • Featuring proprietary Comfort Zone Technology™ for virtually pain-free capillary blood sampling
  • Special design features side release button and preset lancet speed to ensure ease-of-use for healthcare professionals and maximum comfort for patients
  • Simply twist off protective sterile cap, position and activate release button
  • Needle point is hidden before use and automatically retracts after use to avoid needlestick injuries and cross infection
  • Single use device designed to ensure highest level of safety for healthcare professionals and patients
Specifications
Packaging Quantity 25/Pk.
Needle Gauge 30G