Learn More
AdipoGen OX40 (human):Fc (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0014C050
Encompass_Preferred
Protein. Human OX40 (aa 29-214) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse OX40L. Purity: >95% (SDS-PAGE). OX40 binds to receptors on T-cells, preventing them from dying and subsequently increasing cytokine production. OX40 has a critical role in the maintenance of an immune response. OX40:Fc was shown to prevent OX40 from reaching the T-cell receptors, thus reducing the T-cell response. Experiments in mice have demonstrated that OX40:Fc can reduce the symptoms associated with the cytokine storm (an immune overreaction) while allowing the immune system to fight off the virus successfully.
Catalog No. 50-205-3084