Learn More
ACROBiosystems Cynomolgus / Rhesus macaque OX40 Ligand Protein, His Tag, active trimer (MALS verified)
Catalog No. 502331636
Encompass_Preferred
Cynomolgus / Rhesus macaque OX40 Ligand Protein, His Tag (OXL-C5243) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # A0A1D5QQH7 ).
Catalog No. 50-233-1636 Supplier ACROBiosystems Supplier No. OXLC5243100UG
Cynomolgus / Rhesus macaque OX40 Ligand Protein, His Tag (OXL-C5243) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # A0A1D5QQH7 ).