Learn More
Genscript Corporation OX40/TNFRSF4/CD134Biotin Hi
Catalog No. 5000057836
Encompass
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) also known as CD134 and OX40 receptor OX40 is a secondary co-stimulatory immune checkpoint molecule expressed after 24 to 72 hours following activation its ligand OX40L is also not expressed on resting antigen presenting cells but is following their activation
Catalog No. 50-000-57836 Supplier Genscript Corporation Supplier No. Z04578100
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) also known as CD134 and OX40 receptor OX40 is a secondary co-stimulatory immune checkpoint molecule expressed after 24 to 72 hours following activation its ligand OX40L is also not expressed on resting antigen presenting cells but is following their activation