Learn More
Genscript Corporation OX40/TNFRSF4/CD134Biotin Hi
Supplier: Genscript Corporation Z04578500
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) also known as CD134 and OX40 receptor OX40 is a secondary co-stimulatory immune checkpoint molecule expressed after 24 to 72 hours following activation its ligand OX40L is also not expressed on resting antigen presenting cells but is following their activation