Learn More
ACROBiosystems OX40 Protein, Fc Tag, Lyophilized, 1 mg
Catalog No. 501658568
Encompass_Preferred
This recombinant Cynomolgus and Rhesus macaque OX40 (TNFRSF4) protein is expressed in HEK293 cells as a C-terminal human IgG1 Fc-tagged fusion protein. The protein corresponds to amino acids Lys 28 to Ala 214 of the precursor. It is provided in a lyophilized powder format, stabilized with trehalose in a buffer containing Tris, glycine, arginine, and NaCl at pH 7.5.
  • Calculated molecular weight of 46.9 kDa, with migration at 60–66 kDa under reducing conditions on SDS-PAGE due to glycosylation
  • High purity of ≥95% as determined by SDS-PAGE
  • Endotoxin level of <1.0 EU per μg by LAL or rFC method
  • Suitable for research applications requiring validated OX40 interactions
  • Long-term stability of 12 months at -20°C or lower in lyophilized state
Catalog No. 50-165-8568 Supplier ACROBiosystems Supplier No. OX0C52511MG
This recombinant Cynomolgus and Rhesus macaque OX40 (TNFRSF4) protein is expressed in HEK293 cells as a C-terminal human IgG1 Fc-tagged fusion protein. The protein corresponds to amino acids Lys 28 to Ala 214 of the precursor. It is provided in a lyophilized powder format, stabilized with trehalose in a buffer containing Tris, glycine, arginine, and NaCl at pH 7.5.
  • Calculated molecular weight of 46.9 kDa, with migration at 60–66 kDa under reducing conditions on SDS-PAGE due to glycosylation
  • High purity of ≥95% as determined by SDS-PAGE
  • Endotoxin level of <1.0 EU per μg by LAL or rFC method
  • Suitable for research applications requiring validated OX40 interactions
  • Long-term stability of 12 months at -20°C or lower in lyophilized state