Learn More
ACROBiosystems Recombinant Protein;Human OX40 / TNFRSF4 / CD134 Protein, Fc Tag;HEK293;1MG;OX0,TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L,OX-40
Catalog No. 501658528
Encompass_Preferred
Recombinant Protein;Human OX40 / TNFRSF4 / CD134 Protein, Fc Tag;1MG;Human OX40, Fc Tag (OX0-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # NP_003318).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.8 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40 Ligand, His Tag (Cat. No. OXL-H52Q8) at 5 μg/mL (100 μl/well) can bind Human OX40, Fc Tag (Cat. No. OX0-H5255 ) with a linear range of 0.2-5 ng/mL (QC tested).;FACS assay shows that recombinant Human OX40, Fc Tag (Cat. No. OX0-H5255) can bind to 293T cell overexpressing human OX40L. The concentration of OX40 used is 0.03 μg/ml (Routinely tested).
Catalog No. 50-165-8528 Supplier ACROBiosystems Supplier No. OX0H52551MG
Recombinant Protein;Human OX40 / TNFRSF4 / CD134 Protein, Fc Tag;1MG;Human OX40, Fc Tag (OX0-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # NP_003318).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.8 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40 Ligand, His Tag (Cat. No. OXL-H52Q8) at 5 μg/mL (100 μl/well) can bind Human OX40, Fc Tag (Cat. No. OX0-H5255 ) with a linear range of 0.2-5 ng/mL (QC tested).;FACS assay shows that recombinant Human OX40, Fc Tag (Cat. No. OX0-H5255) can bind to 293T cell overexpressing human OX40L. The concentration of OX40 used is 0.03 μg/ml (Routinely tested).