Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse OX40 / TNFRSF4 / CD134 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;OX0,TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L,OX-40
Supplier:  ACROBiosystems OX0M82E525UG
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse OX40 / TNFRSF4 / CD134 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Mouse OX40, His Tag, Avi Tag (OX0-M82E5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession # P47741-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 25.0 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse OX40 Ligand, His Tag (Cat. No. TN4-M5241) at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse OX40, His Tag, Avi Tag (Cat. No. OX0-M82E5) with a linear range of 0.6-10 ng/mL (QC tested).
Catalog No. 50-165-7481