Learn More
Enzo Life Sciences OX40L Recombinant monoclonal antibody (R4930) (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5560200
Encompass_Preferred
Clone: R4930 (Oxelumab). Isotype: IgG1κ. Immunogen: Human OX40L. Species reactivity: Human. Applications: Flow Cytometry, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3082