Learn More
ACROBiosystems ACROBiosystems Biotinylated Human OX40 Ligand / TNFSF4 Protein, Avitag™,Fc Tag
Catalog No. 501957125
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human OX40 Ligand, Avitag,Fc Tag (OXL-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # P23510-1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 48.7 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40, Fc Tag (Cat. No. OX0-H5255) at 1 μg/mL (100 μL/well) can bind Biotinylated Human OX40 Ligand, Avitag,Fc Tag (Cat. No. OXL-H82F4) with a linear range of 0.5-16 ng/mL (QC tested).
Catalog No. 50-195-7125 Supplier ACROBiosystems Supplier No. OXLH82F4200UG
Recombinant Protein;200UG;Biotinylated Human OX40 Ligand, Avitag,Fc Tag (OXL-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # P23510-1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 48.7 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40, Fc Tag (Cat. No. OX0-H5255) at 1 μg/mL (100 μL/well) can bind Biotinylated Human OX40 Ligand, Avitag,Fc Tag (Cat. No. OXL-H82F4) with a linear range of 0.5-16 ng/mL (QC tested).