Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque OX40 Ligand / TNFSF4 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;OXL,OX40L,TNFSF4,CD252,Glycoprotein Gp34,TXGP1
Supplier:  ACROBiosystems OXLR52591MG
Encompass_Preferred
Recombinant Protein;Rhesus macaque OX40 Ligand / TNFSF4 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Rhesus macaque OX40 Ligand, Mouse IgG2a Fc Tag, low endotoxin (OXL-R5259) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # F7FL80-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 42.5 kDa. The protein migrates as 47-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque OX40, His Tag (Cat. No. OX0-C5220) at 2 μg/mL (100 μL/well) can bind Rhesus macaque OX40 Ligand, Mouse IgG2a Fc Tag (Cat. No. OXL-R5259) with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-165-8068