Learn More
Medchemexpress LLC Oxaceprol | 33996-33-7 | 99.7% | 173.17 | 100 G
SDP
Catalog No. 5000387061
Encompass_Preferred
Oxaceprol, an orally active derivative of L-proline, possesses distinct anti-inflammatory activity. It is typically used for the research of osteoarthritis and rheumatoid arthritis. This product is for research use only.
  • Reduces surface area and wet weight of cotton pellet granulomas
  • Inhibits secondary lesions in ears and tail
  • Inhibits neutrophil infiltration into joints
  • Produces comparable inhibition of hyperalgesia to indomethacin
Catalog No. 50-003-87061 Supplier Medchemexpress LLC Supplier No. HY17490100G
Oxaceprol, an orally active derivative of L-proline, possesses distinct anti-inflammatory activity. It is typically used for the research of osteoarthritis and rheumatoid arthritis. This product is for research use only.
  • Reduces surface area and wet weight of cotton pellet granulomas
  • Inhibits secondary lesions in ears and tail
  • Inhibits neutrophil infiltration into joints
  • Produces comparable inhibition of hyperalgesia to indomethacin