Learn More
Medchemexpress LLC Oxaceprol | 33996-33-7 | 99.7% | 173.17 | 25 G
SDP
Catalog No. 5000384400
Encompass_Preferred
Oxaceprol is an orally active derivative of L-proline, possessing distinct anti-inflammatory activity. It is primarily used for research on osteoarthritis and rheumatoid arthritis. Oxaceprol has been shown to significantly reduce the surface area and wet weight of cotton pellet granulomas in vivo, and to inhibit secondary lesions in the ears and tail in animal models. It does not inhibit prostaglandin synthesis in vitro but markedly inhibits neutrophil infiltration into the joints of rats with adjuvant arthritis.
  • Orally active derivative of L-proline
  • Possesses anti-inflammatory activity
  • Used for research on osteoarthritis and rheumatoid arthritis
  • Reduces cotton pellet granulomas in vivo
  • Inhibits secondary lesions in animal models
  • Does not inhibit prostaglandin synthesis in vitro
  • Markedly inhibits neutrophil infiltration into joints
Catalog No. 50-003-84400 Supplier Medchemexpress LLC Supplier No. 25GHY17490
Oxaceprol is an orally active derivative of L-proline, possessing distinct anti-inflammatory activity. It is primarily used for research on osteoarthritis and rheumatoid arthritis. Oxaceprol has been shown to significantly reduce the surface area and wet weight of cotton pellet granulomas in vivo, and to inhibit secondary lesions in the ears and tail in animal models. It does not inhibit prostaglandin synthesis in vitro but markedly inhibits neutrophil infiltration into the joints of rats with adjuvant arthritis.
  • Orally active derivative of L-proline
  • Possesses anti-inflammatory activity
  • Used for research on osteoarthritis and rheumatoid arthritis
  • Reduces cotton pellet granulomas in vivo
  • Inhibits secondary lesions in animal models
  • Does not inhibit prostaglandin synthesis in vitro
  • Markedly inhibits neutrophil infiltration into joints