Learn More
Medchemexpress LLC Oxacillin-13C6 sodium | 1173-88-2 | 99.0% | 429.37 g/mol | C1313C6H18N3NaO5S | 10 MG
SDP
Catalog No. 5000296740
Encompass_Preferred
Oxacillin-13C6 sodium is the 13C6 stable-isotope labeled form of oxacillin sodium supplied as an analytical standard for research and mass spectrometry applications. It provides an isotopically enriched internal standard for accurate quantitation and method validation.
  • 13C6-labeled oxacillin sodium for isotope-dilution assays.
  • High purity: 99.0%.
  • Supplied in small vial sizes suitable for analytical use, including 10 MG.
  • Provided with supporting documentation: SDS, COA, and data sheet.
  • Intended for research and analytical applications; not for clinical use.
Catalog No. 50-002-96740 Supplier Medchemexpress LLC Supplier No. HYB0925S10MG
Oxacillin-13C6 sodium is the 13C6 stable-isotope labeled form of oxacillin sodium supplied as an analytical standard for research and mass spectrometry applications. It provides an isotopically enriched internal standard for accurate quantitation and method validation.
  • 13C6-labeled oxacillin sodium for isotope-dilution assays.
  • High purity: 99.0%.
  • Supplied in small vial sizes suitable for analytical use, including 10 MG.
  • Provided with supporting documentation: SDS, COA, and data sheet.
  • Intended for research and analytical applications; not for clinical use.