Learn More
Medchemexpress LLC Oxacillin-13C6 (sodium) | 1173-88-2 | 99% | 1 MG
SDP
Catalog No. 5000408482
Encompass_Preferred
Oxacillin-13C6 (sodium) is the 13C6 labeled Oxacillin (sodium), a narrow-spectrum β-lactam antibiotic. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs. However, MCE has not independently confirmed the accuracy of these methods.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08482 Supplier Medchemexpress LLC Supplier No. HYB0925S1MG
Oxacillin-13C6 (sodium) is the 13C6 labeled Oxacillin (sodium), a narrow-spectrum β-lactam antibiotic. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs. However, MCE has not independently confirmed the accuracy of these methods.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs