Learn More
Medchemexpress LLC Oxacillin-13C6 sodium | 1173-88-2 | 99% | 5 MG
SDP
Catalog No. 5000408658
Encompass_Preferred
Oxacillin-13C6 sodium is the 13C6 labeled Oxacillin sodium salt, which is a narrow-spectrum β-lactam antibiotic of the penicillin class. This compound is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08658 Supplier Medchemexpress LLC Supplier No. HYB0925S5MG
Oxacillin-13C6 sodium is the 13C6 labeled Oxacillin sodium salt, which is a narrow-spectrum β-lactam antibiotic of the penicillin class. This compound is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS